Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Methyl Tetracosanoate, Fatty Acid Methyl Ester, >99% Purity, 100 mg

Catalog No. 502706072
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6072 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6072 Supplier Abcam Supplier No. AB143876100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly pure fatty acid methyl ester with demonstrated antidiabetic activity in vitro.

  • Molecular Weight: 382.7 Da
  • Purity: >99%
  • Description: Fatty acid methyl ester
  • Biological Activity: Antidiabetic activity; active in vitro
  • Storage buffer: Constituents: 100% Methyl tetracosanoate

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Substrate Properties Lipid-based Substrate
Target Molecule Methyl Tetracosanoate
Format Vial
Content And Storage Store at ambient temperature.
Buffer 100% Methyl tetracosanoate
Formulation 100% Methyl tetracosanoate
Quantity 100 mg
For Use With (Application) Antidiabetic research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.