Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Thiosalicylate 98.0+%, TCI America™
SDP

Catalog No. M188125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M188125G 25 g
M18815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M188125G Supplier TCI America Supplier No. M188125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4892-02-8
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00060678
InChI Key BAQGCWNPCFABAY-UHFFFAOYSA-N
Synonym methyl thiosalicylate, methyl 2-mercaptobenzoate, methyl o-mercaptobenzoate, usaf ma-15, benzoic acid, 2-mercapto-, methyl ester, methyl-o-mercaptobenzoate, methyl-2-mercaptobenzoate, benzoic acid, o-mercapto-, methyl ester, 2-methoxycarbonyl thiophenol, 2-mercaptobenzoic acid methyl ester
PubChem CID 21009
IUPAC Name methyl 2-sulfanylbenzoate
SMILES COC(=O)C1=CC=CC=C1S

Specifications

Color Yellow
Boiling Point 123°C
Quantity 25 g
Formula Weight 168.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Thiosalicylate
RTECSNumber DH3500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.