Learn More
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%
Description
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.
Solubility
Insoluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Solubility Information | Insoluble in water. |
| IUPAC Name | methyl 3-aminocyclohexane-1-carboxylate hydrochloride |
| Quantity | 1 g |
| Formula Weight | 193.67 |
| Percent Purity | 97% |
| Chemical Name or Material | Methyl trans-3-aminocyclohexanecarboxylate hydrochloride |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.