Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl trans-4-(Aminomethyl)cyclohexanecarboxylate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M198825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M198825G 25 g
M19885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M198825G Supplier TCI America Supplier No. M198825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29275-88-5
Molecular Formula C9H18ClNO2
Molecular Weight (g/mol) 207.698
MDL Number MFCD00797465
InChI Key PZPSRTWFJCGJLP-UHFFFAOYSA-N
Synonym trans-4-(Aminomethyl)cyclohexanecarboxylic Acid Methyl Ester Hydrochloride
PubChem CID 12721445
IUPAC Name methyl 4-(aminomethyl)cyclohexane-1-carboxylate;hydrochloride
SMILES COC(=O)C1CCC(CC1)CN.Cl

Specifications

Quantity 25 g
Formula Weight 207.70
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl trans-4-(Aminomethyl)cyclohexanecarboxylate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.