Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl trans-4-Hydroxycyclohexanecarboxylate 98.0+%, TCI America™
SDP

Catalog No. M19911G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19911G 1 g
M19915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19911G Supplier TCI America Supplier No. M19911G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6125-57-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00729087,MFCD20441962,MFCD20441963
InChI Key HYDYVXROZHFTGB-UHFFFAOYSA-N
Synonym trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester
PubChem CID 87117
IUPAC Name methyl 4-hydroxycyclohexane-1-carboxylate
SMILES COC(=O)C1CCC(O)CC1

Specifications

Color Yellow
Boiling Point 125°C
Quantity 1 g
Formula Weight 158.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl trans-4-Hydroxycyclohexanecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.