Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl trans-p-Coumarate 98.0+%, TCI America™
SDP

Catalog No. M225925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M225925G 25 g
M22595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M225925G Supplier TCI America Supplier No. M225925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19367-38-5
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00157167
InChI Key NITWSHWHQAQBAW-UHFFFAOYSA-N
Synonym trans-p-Coumaric Acid Methyl Ester, trans-4-Hydroxycinnamic Acid Methyl Ester, Methyl trans-4-Hydroxycinnamate
PubChem CID 5319562
ChEBI CHEBI:86904
IUPAC Name methyl 3-(4-hydroxyphenyl)prop-2-enoate
SMILES COC(=O)C=CC1=CC=C(O)C=C1

Specifications

Melting Point 139°C
Quantity 25 g
Formula Weight 178.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl trans-p-Coumarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.