Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl trans,trans-9,12-Octadecadienoate 98.0+%, TCI America™
SDP

Catalog No. M2307100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2307100MG 100 mg
M2307500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2307100MG Supplier TCI America Supplier No. M2307100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2566-97-4
Molecular Formula C19H34O2
Molecular Weight (g/mol) 294.48
MDL Number MFCD00069997
InChI Key WTTJVINHCBCLGX-ZDVGBALWSA-N
Synonym methyl linolelaidate, methyl 9e,12e-octadeca-9,12-dienoate, methyl trans,trans-9,12-octadecadienoate, unii-jrb7acq5nw, methyl linoleate hydroperoxide, linolelaidic acid, methyl ester, methyl octadeca-9,12-dienoate, jrb7acq5nw, 9,12-octadecadienoic acid, methyl ester, e,e, linolelaidic acid methyl ester
PubChem CID 5362793
IUPAC Name methyl (9E,12E)-octadeca-9,12-dienoate
SMILES CCCCC\C=C\C\C=C\CCCCCCCC(=O)OC

Specifications

Color Colorless
Boiling Point 152°C
Quantity 100 mg
Formula Weight 294.48
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl trans,trans-9,12-Octadecadienoate
EINECSNumber (2)-0798&(2)-1085
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.