Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Trichloroacetate 96.0+%, TCI America™
SDP

Catalog No. T101325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T101325G 25 g
T1013500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T101325G Supplier TCI America Supplier No. T101325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 598-99-2
Molecular Formula C3H3Cl3O2
Molecular Weight (g/mol) 177.405
MDL Number MFCD00000794
InChI Key VHFUHRXYRYWELT-UHFFFAOYSA-N
Synonym Trichloroacetic Acid Methyl Ester
PubChem CID 11739
IUPAC Name methyl 2,2,2-trichloroacetate
SMILES COC(=O)C(Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 154°C
Quantity 25 g
UN Number 2533
Formula Weight 177.41
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Trichloroacetate
RTECSNumber AJ8380000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.