Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl tridecanoate, 97%

Catalog No. AC438200050
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438200050 5 g
AC438200010 1 g
AC438200100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC438200050 Supplier Thermo Scientific Chemicals Supplier No. 438200050
Add to Cart

CAS: 1731-88-0 | C14H28O2 | 228.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1731-88-0
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.38
InChI Key JNDDPBOKWCBQSM-UHFFFAOYSA-N
Synonym tridecanoic acid, methyl ester, tridecanoic acid methyl ester, methyl n-tridecanoate, unii-o2h463ring, o2h463ring, tridecanoic acid,methyl ester, n-tridecanoic acid methyl ester, methyl tridecylate, methyltridecanoate, tridecanoic acid methyl
PubChem CID 15608
IUPAC Name methyl tridecanoate
SMILES CCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 7°C
Color Colorless
Density 0.8600g/mL
Boiling Point 130°C to 132°C (4.0 Torr)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.86
Formula Weight 228.38
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl tridecanoate
Show More Show Less
EINECSNumber 217-054-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.