Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Methyl Violet Paper Test Strips
SCI_ED

Catalog No. S80848 Shop All Eisco Products
Change view
Click to view available options
pH Range:
0.2 to 2.0
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
S80848 0.2 to 2.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S80848 Supplier Eisco™ Supplier No. METVIOPK12
Add to Cart
Add to Cart

A quick acting, narrow range pH test paper that transitions from yellow at pH 0.2 to violet at pH 2.0

  • Methyl Violet Test Paper
  • Narrow range pH
  • Transitions from yellow at pH 0.2 to violet at pH 2.0
  • Dip test strip in solution for 1-2 seconds and remove
  • No color chart necessary
  • Grasp test strip with fingertips
  • 100 Strips/Vial, 12 Vials/Pack

Specifications

Color Indicator Yellow to Violet
pH Range 0.2 to 2.0
Quantity 100/Pk., 12 Pk./Cs
Test Time 1 to 2 sec.
Type Methyl Violet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.