Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Methylate Reagent (DMFDMA)

Catalog No. PI49350 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With:
GC, Columns 0.53mm I.D., 0.8 mm Ferrule I.D.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
PI49350 GC, Columns 0.53mm I.D., 0.8 mm Ferrule I.D.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI49350 Supplier Thermo Scientific™ Supplier No. TS49350

missing translation for 'validMainframeMsg'

For easy, effective preparation of methyl esters from fatty acids and amino acids use Thermo Scientific™ Methylate Reagent (DMFDMA).

For easy, effective preparation of methyl esters from fatty acids and amino acids use Thermo Scientific™ Methylate Reagent (DMFDMA). Reactions are typically complete upon dissolution (except for long-chain solid acids) and quantitative yields are obtained when reagent and sample are injected without prior mixing. No water washing, extraction, or concentration of derivatives is required and no water is formed. Methylate Reagent (DMFDMA) contains 2 mEq/mL in pyridine and is supplied in ready-to-use Thermo Scientific™ Hypo-Vial™ Sample Storage Vials.

Advantages for preparation of methyl esters for gas chromatography:

  • Speed: the reaction is complete upon dissolution (except long chain solid acids)
  • No water washing, extraction or concentration of derivatives required
  • No water formed
  • Quantitation: quantitative yields are obtained when the reagent and sample are injected without prior mixing
  • Convenient: ready-to-use reagent contains 2mEq/mL pyridine

Ordering Alerts:

Packaged in Hypo-Vial Sample Storage Vial.

TRUSTED_SUSTAINABILITY

Specifications

For Use With GC, Columns 0.53mm I.D., 0.8 mm Ferrule I.D.
Hazards Flammable, Harmful, Toxic
Storage Room Temperature
Standard Components Pyridine, 1,1-Dimethoxy-N,N-dimethyl-methanamine
Standard Concentration 65 - 80, 20 - 25 %

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.