Learn More
Abcam Methylated DNA Quantification Kit, Fluorometric, Quantitative, 48 Tests
Description
- Assay time: 4 hr
- Sensitivity: ≥ 50 pg/well
- Detection method: Fluorescent
- Results type: Quantitative
- Sample types: Saliva, Urine, Plasma, Tissue, Suspension cells, Adherent cells
- Cited in: > 2 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| No. of Reactions | 48 Tests |
| Substrate Properties | Chemical Substrate |
| Target Molecule | 5-Methylcytosine |
| Detection Method | Fluorescence |
| Sample Type | Saliva, Urine, Plasma, Tissue, Suspension Cells, Adherent Cells |
| Quantity | 48 Tests |
| For Use With (Application) | Methylated DNA quantification |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.