Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Methylated DNA Quantification Kit, Fluorometric, Quantitative, 96 Tests
Description
- Detection method: Fluorescent
- Sensitivity: ≥ 50 pg/well
- Assay time: 4 hr
- Sample types: Saliva, Urine, Plasma, Tissue, Suspension cells, Adherent cells
- Results type: Quantitative
- Cited in: 3 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | 5-Methylcytosine (DNA) |
| Format | 96-well Plate Kit |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Microplate Reader |
| No. of Reactions | 96 Tests |
| Sample Type | Saliva, Urine, Plasma, Tissue, Suspension Cells, Adherent Cells |
| Quantity | 96 Tests |
| For Use With (Application) | 5-Methylcytosine quantification |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?