Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Methylated Human Control for MethylEdge™ Bisulfite Conversion System

Catalog No. PRN1231 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN1231 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRN1231 Supplier Promega Supplier No. N1231
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylated Human Control DNA is purified from a human male source. CpG sites in the DNA are enzymatically methylated by M. SssI methyltransferase to provide a high percentage of methylated CpG motifs.

CpG sites in the DNA are enzymatically methylated by M. SssI methyltransferase to provide a high percentage of methylated CpG motifs. Methylated Human Control DNA can be bisulfite-converted with MethylEdge™ Bisulfite Conversion System in parallel with experimental samples to assess conversion efficiency.

Specifications

Content And Storage 2°C to 10°C
Format Tube
Product Type Control DNA
Quantity 5 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.