Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Methylbenzyl acetate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019906 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9906 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9906 Supplier MilliporeSigma Supelco Supplier No. 789751ML

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 93-92-5
Density 1.028 g/mL (at 25°C)
Flash Point 91°C - Closed Cup
Physical Form Neat
Linear Formula CH3 CO2 CH(CH3)C6H5
MDL Number MFCD00008858
Boiling Point 94°C to 95°C (12 mmHg)
Synonym alpha-Methylbenzyl acetate; alpha-Methylbenzenemethanol 1-acetate; 1-Phenylethyl acetate; Gardenol; Styrallyl acetate
Type Analytical Standard
Quantity 1 mL
Formula Weight 164.20
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.