Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcyclopentadiene dimer, tech. 90%, stab. with 200ppm 4-tert-butylcatechol

Catalog No. AAA175960J
Change view
Click to view available options
Quantity:
500 mL
1000 mL
5000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA175960J 5000 mL
AAA17596AP 500 mL
AAA17596AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA175960J Supplier Thermo Scientific Chemicals Supplier No. A175960J
Add to Cart

CAS: 26472-00-4 | C12H16

Methylcyclopentadiene dimer is used to study the competition with ethylene for binding and biological activities. Used in the synthesis of rubbers and resins intended for use in contact with food or drink.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylcyclopentadiene dimer is used to study the competition with ethylene for binding and biological activities. Used in the synthesis of rubbers and resins intended for use in contact with food or drink.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26472-00-4
Molecular Formula C12H16
MDL Number MFCD00082297

Specifications

Melting Point ∼-50°C
Density 0.94
Boiling Point 198°C to 201°C
Flash Point 26°C (79°F)
Refractive Index 1.498
Quantity 5000 mL
UN Number UN3295
Solubility Information Insoluble in water.
Formula Weight 160.26
Percent Purity 90%
Grade Technical
Chemical Name or Material Methylcyclopentadiene dimer, stabilized with 200ppm 4-tert-butylcatechol
Show More Show Less
Hazard Category H226-H351
Hazard Statement GHS H Statement
H340-H350-H226
May cause genetic defects.
May cause cancer.
Flammable liquid and vapour.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P280-P308+P313-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 247-724-5
RTECSNumber PC1075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.