Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyldiphenylphosphine, 99%

Catalog No. AAH2715909
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2715909 10 g
AAH2715903 1 g
AAH2715918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2715909 Supplier Thermo Scientific Chemicals Supplier No. H2715909
Add to Cart

CAS: 1486-28-8 | C13H13P | 200.22 g/mol

Methyldiphenylphosphine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyldiphenylphosphine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1486-28-8
Molecular Formula C13H13P
Molecular Weight (g/mol) 200.22
MDL Number MFCD00008508
InChI Key UJNZOIKQAUQOCN-UHFFFAOYSA-N
Synonym methyldiphenylphosphine, diphenylmethylphosphine, methyl diphenyl phosphine, phosphine, methyldiphenyl, methyldiphenylphsphine, diphenylphosphinomethane, methyldiphenylphoshine, pubchem6421, methyl diphenylphosphine, methyldi-phenylphosphine
PubChem CID 73879
IUPAC Name methyl(diphenyl)phosphane
SMILES CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 1.064
Boiling Point 160°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 200.21
Percent Purity 99%
Chemical Name or Material Methyldiphenylphosphine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 216-065-5
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.