Learn More
Description
Includes
- Collection Tubes
- ME Spin Columns
- ME Elution Buffer
- ME Desulphonation Buffer
- ME Wash Buffer
- ME Binding Buffer
- ME Conversion Reagent
- Rapid Bisulfite Conversion with Minimal DNA Fragmentation
- High-efficiency DNA conversion from as little as 100pg input DNA
- Rapid protocol saves time compared to other conversion systems
- Robust conversion of DNA with reduced DNA fragmentation
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Format | Kit |
| Isolation Technology | Silica Spin Column |
| No. of Reactions | 50 |
| Product Type | Bisulfite Conversion Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.