Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue, 1% AQ, Vion Biosciences
SCI_ED

Catalog No. S02490 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02490 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02490 Supplier Aldon Corporation Supplier No. VNMM0440500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stains cells an intense blue for ease of visualization in histological specimens with 1% aqueous methylene blue.

  • Stains cells an intense blue for ease of visualization in histological specimens with 1% aqueous methylene blue.

Chemical Identifiers

CAS 61-73-4 , 7732-18-5 , 532-32-1
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.85
MDL Number MFCD00150008
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
Synonym Methylene Blue Chloride, Water Solution
PubChem CID Data not available.
ChEBI CHEBI:6872
IUPAC Name 3,7-bis(dimethylamino)-5λ⁴-phenothiazin-5-ylium chloride
SMILES [Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

Melting Point ~0°C (32°F in water)
Boiling Point ~100°C (212°F in water)
CAS Min % 1.00%, 98.80%, 0.20%
Concentration 1%
Dye Content 1%
CAS Max % 98.80%, 1.00%, 0.20%
Infrared Spectrum Yes
Lambda Max 665 nm
Odor No odor.
Packaging HDPE Bottle
Specific Gravity 1.1 g/mL
Assay Percent Range 1.00%
Vapor Pressure 14 mm Hg in water)
Certificate of Analysis Yes
UN Number Not regulated.
Decomposition Information Data not available.
Density 1.1 g/mL
pH Color Changes Data not available.
Solvent or Matrix Water
Color Blue
Solubility Information Complete in water.
pH Data not available.
Quantity 500 mL
Flash Point Data not available.
Physical Form Data not available.
Percent Purity 1%
Grade Data not available.
Concentration or Composition (by Analyte or Components) 1% Aqueous
Chemical Name or Material Methylene Blue Chloride
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Acute Toxicity, Oral
ShelfLife 3 years
DOTInformation Not regulated.
EINECSNumber 200-515-2
RTECSNumber SO5600000
Recommended Storage Store in a cool, well-ventilated area away from incompatible substances.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.