Learn More
Novus Biologicals™ Methylene Blue

Description
Biological Activity:Biological stain and redox indicator. Inhibits tau filament formation (IC50 = 1.9µM). Also inhibits soluble guanylyl cyclase.
Storage: Store at 4°C.
Note: Inhibitors are guranteed for 1 year from date of receipt.
Specifications
Specifications
| Description | CAS: 61-73-4 |
| Formulation | C16H18ClN3S |
| For Use With (Application) | Inhibits apoptosis |
| Quantity | 50 mg |
| Product Type | Inhibitors |
| Molecular Weight (g/mol) | 319.85g/mol |
| Solubility | Soluble to 10 mM in water, 10 mM in DMSO |
| Content And Storage | 4°C |
| Target | Methylene Blue |
| Color | Green |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.