Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Methylene Blue
SCI_ED

Catalog No. S25429 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25429 10 g Solid
S25429A 25 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25429
Biological stain

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7220-79-3 , 7732-18-5
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym Methylene Blue Chloride, Basic Blue 9
PubChem CID 104827
IUPAC Name 3,7-bis(dimethylamino)-5λ⁴-phenothiazin-5-ylium trihydrate chloride
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

Assay Percent Range 1% w/v
Linear Formula C16H18ClN3S·3H2O
Formula Weight 373.9
Packaging Solid
Density ∽1.00g/mL at 20°C
Color Green
Quantity 10 g
Chemical Name or Material Methylene Blue
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.