Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue, Anhydrous, Spectrum™ Chemical
SDP

Catalog No. 18621137 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-137 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-621-137 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M32715GM

missing translation for 'validMainframeMsg'

Synonym: Basic Blue 9, Methylthioninium Chloride, Solvent Blue 8, Swiss Blue, Ehrlichs Reagent III

Methylene Blue is a heterocyclic aromatic solid that is an odorless powder that has a dark green tinge and turns to blue when mixed with water. It is used in aquaculture and by tropical fish hobbyists as a treatment for fungal infections and has many experimental pharmaceutical drug applications. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 61-73-4

Specifications

Packaging Amber Glass
Color Dark Green
Quantity 5 g
Chemical Name or Material Methylene Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.