Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003392 Shop All Avantor Products
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-392 25 g Wide Mouth Amber Glass Bottle
02-003-394 500 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-003-392 Supplier Avantor Supplier No. Q47303

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym C.I. 52015
PubChem CID 104827
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
Formula Weight 373.91
Grade BAKER ANALYZED™ Reagent
Packaging Wide Mouth Amber Glass Bottle
Quantity 25 g
Chemical Name or Material Methylene Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.