Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue Loeffler Formulation, Loeffler's Alkaline Methylene Blue, Ricca Chemical
SDP

Catalog No. 48804 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
120 mL
500 mL
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4880-4 120 mL Natural Poly Bottle
R4880000100 100 mL Natural Poly Bottle
4880-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 4880-4 Supplier Ricca Chemical Company Supplier No. 48804
Add to Cart
Add to Cart

For staining and differentiation of acid fast organisms in smears

Specifications

Chemical Name or Material METHYLENE BLUE
CAS 7732-18-5, 64-17-5, 7220-79-3, 1310-58-3
CAS Min % 79.0, 18.91, 0.24, 0.01
CAS Max % 82.0, 19.68, 0.25, 0.01
Grade Laboratory
Packaging Natural Poly Bottle
Color Blue
Quantity 120 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.