Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue Reagent, for Surfactant Analysis, Ricca Chemical
SDP

Catalog No. 490016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4900-16 500 mL Natural Poly Bottle
4900-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4900-16 Supplier Ricca Chemical Company Supplier No. 490016
Add to Cart
Add to Cart

For Analysis of Surfactants

Specifications

Chemical Name or Material Methylene Blue Reagent
CAS 7732-18-5, 10049-21-5, 7664-93-9, 7732-18-5
Grade Laboratory
Packaging Natural Poly Bottle
Color Dark Blue
Quantity 500 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.