Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue, Sigma-Aldrich

Catalog No. 01258814 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-814 25 g
01-259-081 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-814 Supplier MilliporeSigma Supelco Supplier No. M914025G
Add to Cart
Add to Cart

Certified for use in Mallory's phloxine-methylene blue stain; Levine and Black stain for bacteria in milk; Wright stain for blood smears.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methylene Blue
CAS 122965-43-9
Molecular Weight (g/mol) 319.85 g/mol
Grade Certified Biological Stain
Color Dark Green
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.