Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue Trihydrate, Zinc Free, MP Biomedicals

Catalog No. ICN19399805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19399805 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19399805 Supplier MP Biomedicals, Inc Supplier No. 0219399805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An ethidium bromide alternative for the visualization of nucleic acids in gels.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-73-4
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.851
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
Synonym Methylthionine chloride, C.I. 52015
PubChem CID 6099
ChEBI CHEBI:6872
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]

Specifications

Color Dark Blue
Quantity 5 g
Formula Weight 373.9
Grade Not Available
Chemical Name or Material Methylene Blue, An ethidium bromide alternative for the visualization of nucleic acids in gels
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.