Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, FCC, 99%, Spectrum™ Chemical
SDP

Catalog No. 18604501 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Steel Can
Steel Drum
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-501 1 L Amber Glass Bottle
18-604-500 100 mL Amber Glass Bottle
18-604-506 500 mL Amber Glass Bottle
18-604-507 6 x 500 mL Amber Glass Bottle
18-604-502 4 L Amber Glass Bottle
18-604-503 6 x 1 L Amber Glass Bottle
18-604-504 4 x 4 L Amber Glass Bottle
18-604-505 20 L Steel Can
18-604-508 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 18-604-501 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M11721LT
Add to Cart
Add to Cart
This item is not returnable. View return policy

M1172, 75-09-2, CH2Cl2

Spectrum™ Chemical Methylene Chloride, FCC is used to decaffeinate coffee and tea as well as to prepare extracts of hops and other flavorings. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Acidity 10 mg/kg
CAS Max % 100%
Quantity 1 L
Packaging Amber Glass Bottle
Specific Gravity 1.318-1.323
Water 0.02%
UN Number UN1593
Boiling Point 39.8°C
Percent Purity 99%
Grade FCC
Chemical Name or Material Methylene Chloride
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.