Learn More
Methylene Chloride, FCC, 99%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Methylene Chloride, FCC is used to decaffeinate coffee and tea as well as to prepare extracts of hops and other flavorings. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| Acidity | 10 mg/kg |
| CAS Max % | 100% |
| Quantity | 6 x 500 mL |
| Packaging | Amber Glass Bottle |
| Specific Gravity | 1.318-1.323 |
| Water | 0.02% |
| UN Number | UN1593 |
| Boiling Point | 39.8°C |
| Percent Purity | 99% |
| Grade | FCC |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.