Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Glycol Bis(2,3-dibromopropyl) Ether 70.0+%, TCI America™
SDP

Catalog No. M031325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M031325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M031325ML Supplier TCI America Supplier No. M031325ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34446-11-2
Molecular Formula C7H12Br4O2
Molecular Weight (g/mol) 447.787
MDL Number MFCD00059904
InChI Key DUQWOWBBMLWHPB-UHFFFAOYSA-N
Synonym Formaldehyde Bis(2,3-dibromopropyl Acetal)
PubChem CID 12585580
IUPAC Name 1,2-dibromo-3-(2,3-dibromopropoxymethoxy)propane
SMILES C(C(CBr)Br)OCOCC(CBr)Br

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 mL
Formula Weight 447.79
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material Methylene Glycol Bis(2,3-dibromopropyl) Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.