Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene green, MP Biomedicals™

Catalog No. ICN15264405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15264405 5 g
ICN15264425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15264405 Supplier MP Biomedicals, Inc Supplier No. 0215264405
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2679-01-8 | C16H17ClN4O2S | 364.85 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2679-01-8
Molecular Formula C16H17ClN4O2S
Molecular Weight (g/mol) 364.85
InChI Key YYGBVRCTHASBKD-UHFFFAOYSA-M
Synonym C.I. 52020, 3, 7-Bis(Dimethylamino)-4-Nitro-Phenothiazin-5-Ium Chloride
ChEBI CHEBI:87675

Specifications

Color Black
Quantity 5 g
Physical Form Powder
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C(=C3S2)[N+](=O)[O-].[Cl-]
IUPAC Name [7-(dimethylamino)-4-nitrophenothiazin-3-ylidene]-dimethylazanium;chloride
PubChem CID 75889
Formula Weight 364.86
Percent Purity >75.0%
Grade Histological
Chemical Name or Material Methylene green
EINECSNumber 220-231-2
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.