Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylenebis(thioglycolic Acid) 96.0+%, TCI America™
SDP

Catalog No. M021825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M021825G 25 g
M02185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M021825G Supplier TCI America Supplier No. M021825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2068-24-8
Molecular Formula C5H8O4S2
Molecular Weight (g/mol) 196.235
MDL Number MFCD00135949
InChI Key CZEVAMMETWVBOD-UHFFFAOYSA-N
Synonym methylenedithiodiacetic acid, methylenebisthioglycolic acid, acetic acid, 2,2'-methylenebis thio bis, acetic acid, methylenedithio di, 2,2'-methylenebis thio diacetic acid, methylenebis-thioglycollic acid, 2,2'-methylenebis sulfanediyl diacetic acid, 2-carboxymethyl thio methyl thio acetic acid, carboxymethyl sulfanyl methyl sulfanyl acetic acid
PubChem CID 74949
IUPAC Name 2-(carboxymethylsulfanylmethylsulfanyl)acetic acid
SMILES C(C(=O)O)SCSCC(=O)O

Specifications

Melting Point 129°C
Quantity 25 g
Formula Weight 196.24
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methylenebis(thioglycolic Acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.