Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl vanillate, 99%

Catalog No. AC160300250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160300250 25 g Glass bottle
AC160300050 5 g Glass bottle
AC160301000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC160300250 Supplier Thermo Scientific Chemicals Supplier No. 160300250
Only null left
Add to Cart

CAS: 3943-74-6 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3943-74-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00008438
InChI Key BVWTXUYLKBHMOX-UHFFFAOYSA-N
Synonym methyl vanillate, methyl 3-methoxy-4-hydroxybenzoate, vanillic acid, methyl ester, 4-hydroxy-3-methoxybenzoic acid methyl ester, methylvanillate, benzoic acid, 4-hydroxy-3-methoxy-, methyl ester, vanillic acid methyl ester, unii-2hxg8qso3d, 2hxg8qso3d, vxx
PubChem CID 19844
ChEBI CHEBI:46477
IUPAC Name methyl 4-hydroxy-3-methoxybenzoate
SMILES COC1=C(C=CC(=C1)C(=O)OC)O

Specifications

Melting Point 64.0°C to 67.0°C
Color Beige to Yellow
Boiling Point 285.0°C to 287.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)CO2CH3
Quantity 25 g
Beilstein 10, 396
Formula Weight 182.18
Percent Purity 99%
Physical Form Powder or Granules
Chemical Name or Material Methyl vanillate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 223-525-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.