Learn More
Cell Biolabs Inc OxiSelect™ Methylglyoxal (MG) Competitive ELISA Kit, 96 assays
Supplier:  Cell Biolabs Inc STA811
Encompass_Preferred
Methylglyoxal (MG) is a common species of advanced glycation end products (AGE). AGE species are formed from the Maillard reaction, also known as glycation, which is a non-enzymatic reaction of reducing carbohydrates with the N-terminus and/or lysine side chains of proteins, phospholipids, and nucleic acids. Upon formation, MG readily forms adducts with proteins. Cell Biolabs' OxiSelect™ Methylglyoxal Competitive ELISA Kit provides rapid detection of MG-protein adducts in cell and tissue lysates, plasma, serum, or purified proteins. Each kit provides sufficient reagents for 96 assays including standard curve and unknown protein samples.
Catalog No. 50-673-727
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.