Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylhydrazine sulfate, 98%

Catalog No. AAL1012206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1012206 5 g
AAL1012214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1012206 Supplier Thermo Scientific Chemicals Supplier No. L1012206
Add to Cart

CAS: 302-15-8 | CH8N2O4S | 144.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-15-8
Molecular Formula CH8N2O4S
Molecular Weight (g/mol) 144.15
MDL Number MFCD00035423
InChI Key KJDJPXUIZYHXEZ-UHFFFAOYSA-N
Synonym methylhydrazine sulfate, methylhydrazine; sulfuric acid, methyl hydrazine sulfate, methyl hydrazine sulphate, hydrazine, methyl-, sulfate, methyl hydrazine; sulfuric acid, ch6n2.h2o4s, methylhydrazine sulphate, dsstox_cid_875, n-methyl hydrazine sulfate
PubChem CID 165286
IUPAC Name methylhydrazine;sulfuric acid
SMILES CN[NH3+].OS([O-])(=O)=O

Specifications

Melting Point 141°C to 143°C
Quantity 5 g
UN Number UN2811
Beilstein 3693575
Merck Index 14,6086
Formula Weight 144.15
Percent Purity 98%
Chemical Name or Material Methylhydrazine sulfate
Hazard Category H301+H311-H330-H351
Hazard Statement GHS H Statement
H301-H311-H331-H351
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P308+P313-P310-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 206-115-4
RTECSNumber MV7750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.