Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyllithium lithium bromide complex, 2.2M sol. in diethylether, AcroSeal™, Thermo Scientific™

Catalog No. AC181295000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC181295000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC181295000 Supplier Thermo Scientific Chemicals Supplier No. 181295000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332360-06-2 , 60-29-7
Molecular Formula CH3Li·LiBr
MDL Number MFCD00044645
Synonym dilithium carbanide bromide, methyllithium lithium bromide complex solution, dilithium 1+ ion bromide methanide

Specifications

Color Colorless to Yellow
Density 0.81g/mL
Quantity 500 mL
Physical Form Solution
Linear Formula CH3Li·LiBr
Formula Weight 108.82
Percent Purity 9.0 to 11.0% (LiBr), 6.0 to 7.0% (LiCH3)
Chemical Name or Material Methyllithium lithium bromide complex
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May cause an allergic skin reaction. Reacts violently with water. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. Handle under inert gas. Protect from moisture. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician.
Recommended Storage Product may develop some turbidity or precipitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.