Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylphosphonic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAA1261914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1261914 25 g
AAA1261903 1 g
AAA1261906 5 g
AAA1261922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1261914 Supplier Thermo Scientific Chemicals Supplier No. A1261914
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Methylphosphonic acid is used to produce methyl-phosphonic acid dichloride by chlorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylphosphonic acid is used to produce methyl-phosphonic acid dichloride by chlorination.

Solubility
Soluble in water and alcohol.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 993-13-5
Molecular Formula CH5O3P
Molecular Weight (g/mol) 96.02
MDL Number MFCD00002137
InChI Key YACKEPLHDIMKIO-UHFFFAOYSA-N
Synonym methanephosphonic acid, phosphonic acid, methyl, methyl phosphonic acid, unii-329w4ym10z, phosphonomethyl group, ester group, phosphonomethyl, acmc-20ake9, d02fxw, methylphosphonic acid
PubChem CID 13818
ChEBI CHEBI:45129
IUPAC Name methylphosphonic acid
SMILES CP(O)(O)=O

Specifications

Melting Point 103°C to 107°C
Quantity 25 g
UN Number UN3261
Beilstein 1739372
Sensitivity Hygroscopic
Solubility Information Soluble in water and alcohol.
Formula Weight 96.03
Percent Purity 98%
Chemical Name or Material Methylphosphonic acid
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 213-607-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.