Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methylphosphonic Dichloride

Catalog No. 22805225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-805-225G 25 g
22-805-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-805-225G Supplier Sigma Aldrich Supplier No. 22805225G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methylphosphonic Dichloride

Specifications

CAS 676-97-1
Melting Point 35°C to 37°C
Density 1.468 g/mL at 25°C
Boiling Point 163°C
Molecular Formula CH3Cl2OP
Linear Formula CH3P(O)Cl2
MDL Number MFCD00002071
Quantity 25 g
Synonym Dichloromethylphosphine oxide; Methanephosphonyl chloride; Methylphosphonodichloridic acid; Methylphosphonyl chloride; Methylphosphonyl dichloride
Molecular Weight (g/mol) 132.91 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.