Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Methylsulfonyl)methanesulfonyl chloride, 98%

Catalog No. aah5038503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5038503 1 g
AAH5038506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5038503 Supplier Thermo Scientific Chemicals Supplier No. H5038503
Only null left
Add to Cart

CAS: 22317-89-1 | C2H5ClO4S2 | 192.628 g/mol

(Methylsulfonyl)methanesulfonyl chloride is used as a chemicals, pharmaceutical intermediates, reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Methylsulfonyl)methanesulfonyl chloride is used as a chemicals, pharmaceutical intermediates, reagents.

Solubility
Soluble in water (50g/L).

Notes
Moisture Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22317-89-1
Molecular Formula C2H5ClO4S2
Molecular Weight (g/mol) 192.628
MDL Number MFCD00052809
InChI Key DQDYGBSUPOHHMW-UHFFFAOYSA-N
Synonym methylsulfonylmethylsulfonyl chloride, methylsulfonyl-methanesulfonyl chloride, methanesulfonylmethanesulfonyl chloride, methylmethanedisulfonyl chloride, methylsulfonyl methanesulfonyl chloride, acmc-1cql3, methanesulfonyl mesyl chloride, chloro methylsulfonyl methyl sulfone, methanesulfonyl-methanesulfonyl chloride, methanesulfonyl methanesulfonyl chloride
PubChem CID 556269
IUPAC Name methylsulfonylmethanesulfonyl chloride
SMILES CS(=O)(=O)CS(=O)(=O)Cl

Specifications

Melting Point 110°C
Linear Formula ClSO2CH2SO2CH3
Quantity 1 g
UN Number UN3261
Sensitivity Hygroscopic; Moisture sensitive
Solubility Information Soluble in water (50g/L).
Formula Weight 192.63
Percent Purity 98%
Chemical Name or Material (Methylsulfonyl)methanesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.