Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylthymol Blue Sodium Salt 95.0+%, TCI America™
SDP

Catalog No. B047825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B047825G 25 g
B04781G 1 g
B04785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B047825G Supplier TCI America Supplier No. B047825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1945-77-3
Molecular Formula C37H44N2Na4O13S
Molecular Weight (g/mol) 848.779
MDL Number MFCD00148905
InChI Key LGVVZVZPOQJZKT-UHFFFAOYSA-N
PubChem CID 131850582
IUPAC Name 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]

Specifications

Color Black/Green
Synonym 3,3′C-Bis[N,N-bis(carboxymethyl)aminomethyl]thymolsulfonphthalein Sodium Salt
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 844.75
Percent Purity ≥95.0% (HPLC)
Chemical Name or Material Methylthymol Blue Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.