Learn More
Cayman Chemical Methyltransferase Colorimetri
SDP
Supplier:  Cayman Chemical 7001441 EA
Encompass_Preferred
Lipid CP-LC-1074 is an ionizable cationic amino lipid derived from homocysteine a naturally occurring amino acid. This lipid has demonstrated an efficient delivery and high protein expression of different kinds of RNA (mRNA cRNA and saRNA)in vivo. Moreover using a typical LNP formulation (IL/Ch/DOPE/PEG 50/38.5/10/1.5) it has shown to induce specific lung targeting much more efficiently that other commercial alternatives throUGh IV injection with no signs of toxicity.
Catalog No. 50-289-3470
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.