Learn More
Methyltriphenylphosphonium bromide, 98+%
Description
Methyltriphenylphosphonium bromide is used for methylenation through the Wittig reaction. It is utilized in the synthesis of an enyne and 9-isopropenyl -phenanthrene by using sodium amide as reagent. It is a lipophilic molecule and used to deliver molecules to specific cell components. Further, it acts as an antineoplastic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyltriphenylphosphonium bromide is used for methylenation through the Wittig reaction. It is utilized in the synthesis of an enyne and 9-isopropenyl -phenanthrene by using sodium amide as reagent. It is a lipophilic molecule and used to deliver molecules to specific cell components. Further, it acts as an antineoplastic agent.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 231°C to 234°C |
| Assay Percent Range | ≥98% |
| Quantity | 1000 g |
| UN Number | UN2811 |
| Beilstein | 3599467 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water. |
| Formula Weight | 357.24 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Methyltriphenylphosphonium bromide |
Safety and Handling
| Hazard Category | H301-H335-H500 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 217-218-9 |
| TSCA | Yes |
| storageNote1 | Hygroscopic |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.