Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metolazone, MP Biomedicals™

Catalog No. ICN15361780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15361780 100 mg
ICN15361725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15361780 Supplier MP Biomedicals, Inc Supplier No. 0215361780

missing translation for 'validMainframeMsg'

Diuretic; anti-hypertensive. Moderately soluble in water; soluble in ethanol or DMSO (Store at 2° to 8°C)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17560-51-9
Molecular Formula C16H16ClN3O3S
Molecular Weight (g/mol) 365.83
MDL Number MFCD00069304
InChI Key AQCHWTWZEMGIFD-UHFFFAOYNA-N
Synonym metolazone, zaroxolyn, diulo, mykrox, microx, metalozone, metenix, oldren, metolazona, metolazonum
PubChem CID 4170
ChEBI CHEBI:64354
IUPAC Name 7-chloro-2-methyl-3-(2-methylphenyl)-4-oxo-1,2,3,4-tetrahydroquinazoline-6-sulfonamide
SMILES CC1NC2=CC(Cl)=C(C=C2C(=O)N1C1=CC=CC=C1C)S(N)(=O)=O

Specifications

Color White
Quantity 100 mg
Formula Weight 365.83
Physical Form Solid
Chemical Name or Material Metolazone
EINECSNumber 241-539-3
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.