Learn More
CPC Scientific H-Tyr-Gly-Gly-Phe-Met-Arg-Arg-Val-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific METO001B
Encompass
This peptide is the C-terminally amidated octapeptide found in bovine brain and adrenal tissue, corresponds to residues 206-213 of the bovine proenkephalin precursor. It is also related in sequence to the bovine adrenal medulla peptides. So far, it is the only endogenous opioid peptide with high affinity for ? opioid receptors.ONE-LETTER SEQUENCE: YGGFMRRV-NH2MOLECULAR FORMULA: C44H69N15O9S1MOLECULAR WEIGHT:984.19STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [88377-68-8], RESEARCH AREA: PainREFERENCES: H.Matsuo et al., Nature, 305, 721 (1983); E.Weber et al., Proc. Natl. Acad. Sci. USA, 80, 7362 (1983); P.D.Marley et al., Brain Res., 363, 10 (1986); M.Sonders and E.Weber, J. Neurochem., 49, 671 (1987)
Catalog No. 50-210-9726
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.