Learn More
Cytiva (Formerly Pall Lab) Metricel Black PES Membrane Disc Filter - 0.45 m, 47 mm, Gridded (100/pkg)
Catalog No. 502063340 Shop All Cytiva Products
Encompass
Contrasting Membrane for Microbiological Analysis-Dark background provides excellent contrast for counting opaque colonies in microbiology labs-Exclusive dot grid pattern does not enhance or inhibit colony growth-Certified for use in the Membrane Filter (MF) Technique as described in Standard Methods for the Examination of Water and Wastewater, current edition-Sharp contrast between black membrane and white grid line provides guidance while viewing and counting-Available non-sterile or in individual gamma-irradiated packs (S-packs) for critical applications This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3340 Supplier Cytiva Supplier No. 66586
Contrasting Membrane for Microbiological Analysis-Dark background provides excellent contrast for counting opaque colonies in microbiology labs-Exclusive dot grid pattern does not enhance or inhibit colony growth-Certified for use in the Membrane Filter (MF) Technique as described in Standard Methods for the Examination of Water and Wastewater, current edition-Sharp contrast between black membrane and white grid line provides guidance while viewing and counting-Available non-sterile or in individual gamma-irradiated packs (S-packs) for critical applications This product is not approved for use in medical, clinical, surgical or other patient applications.