Learn More
Cytiva Metricel Black Membrane, 0.45 µm, 47 mm, Black, Gridded, Gamma Irradiated, Bulk w/ Parchment Separators (200/pkg)
Catalog No. 502063339 Shop All Cytiva Products
Encompass
Contrasting Membrane for Microbiological Analysis-Dark background provides excellent contrast for counting opaque colonies in microbiology labs-Exclusive dot grid pattern does not enhance or inhibit colony growth-Certified for use in the Membrane Filter (MF) Technique as described in Standard Methods for the Examination of Water and Wastewater, current edition-Sharp contrast between black membrane and white grid line provides guidance while viewing and counting-Available non-sterile or in individual gamma-irradiated packs (S-packs) for critical applications This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3339 Supplier Cytiva Supplier No. 66585
Contrasting Membrane for Microbiological Analysis-Dark background provides excellent contrast for counting opaque colonies in microbiology labs-Exclusive dot grid pattern does not enhance or inhibit colony growth-Certified for use in the Membrane Filter (MF) Technique as described in Standard Methods for the Examination of Water and Wastewater, current edition-Sharp contrast between black membrane and white grid line provides guidance while viewing and counting-Available non-sterile or in individual gamma-irradiated packs (S-packs) for critical applications This product is not approved for use in medical, clinical, surgical or other patient applications.