Learn More
Cytiva Metricel Black Membrane, 0.8 µm, 47 mm, Black, Gridded, Gamma Irradiated, Bulk w/ Parchment Separators (200/pkg)
Catalog No. NC1671347 Shop All Cytiva Products
Encompass
Contrasting Membrane for Microbiological Analysis-Dark background provides excellent contrast for counting opaque colonies in microbiology labs-Exclusive dot grid pattern does not enhance or inhibit colony growth-Certified for use in the Membrane Filter (MF) Technique as described in Standard Methods for the Examination of Water and Wastewater, current edition-Sharp contrast between black membrane and white grid line provides guidance while viewing and counting-Available non-sterile or in individual gamma-irradiated packs (S-packs) for critical applications This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1671347 Supplier Cytiva Supplier No. 66587
Contrasting Membrane for Microbiological Analysis-Dark background provides excellent contrast for counting opaque colonies in microbiology labs-Exclusive dot grid pattern does not enhance or inhibit colony growth-Certified for use in the Membrane Filter (MF) Technique as described in Standard Methods for the Examination of Water and Wastewater, current edition-Sharp contrast between black membrane and white grid line provides guidance while viewing and counting-Available non-sterile or in individual gamma-irradiated packs (S-packs) for critical applications This product is not approved for use in medical, clinical, surgical or other patient applications.