Learn More
Metro™ High-Density qwikTRAK™ Mobile Unit Kits
GSA_VA
Catalog No. 17823712 Shop All Metro Products
Change view
Click to view available options
For Use With (Equipment):
Metro MetroMax i and Q Shelving System
Metro Super Erecta, Super Adjustable Shelving System
Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System
Material:
Polymer
Stainless Steel
Steel
Weight (English):
18.5 lb.
25 lb.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material Weight (English)
17-823-712 Metro Super Erecta, Super Adjustable Shelving System Stainless Steel 25 lb.
17-823-710 Metro Super Erecta, Super Adjustable Shelving System Steel 25 lb.
17-823-711 Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System Steel 25 lb.
17-823-714 Metro MetroMax i and Q Shelving System Polymer 18.5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-823-712 Supplier Metro™ Supplier No. BTMS
Add to Cart
Add to Cart
Kit provides components for shelving units that fit on tracks for added mobility
  • For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax iQ™ shelving systems
  • Kit includes four 74 in. (188cm) high mobile posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Order Info
Order one kit per mobile system. Shelves are sold separately.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Mobile Unit Kit
For Use With (Equipment) Metro Super Erecta, Super Adjustable Shelving System
Includes Four 74 in. high mobile posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Weight (English) 25 lb.
Weight (Metric) 11.4 kg
Material Stainless Steel