Learn More
Metro™ MetroMax i™ Top-Track Mobile Unit Kits
Catalog No. 17820339 Shop All Metro Products
Change view
Click to view available options
Width (English) Shelf:
18 in.
24 in.
Includes:
Posts, Chrome-plated rigid casters, caster channel, bumpers and roller assemblies
Posts, Stainless Steel rigid casters, caster channel, bumpers and roller assemblies
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Shelf Includes
17-820-339 24 in. Posts, Chrome-plated rigid casters, caster channel, bumpers and roller assemblies
17-820-337 18 in. Posts, Chrome-plated rigid casters, caster channel, bumpers and roller assemblies
17-820-338 18 in. Posts, Stainless Steel rigid casters, caster channel, bumpers and roller assemblies
17-820-340 24 in. Posts, Stainless Steel rigid casters, caster channel, bumpers and roller assemblies
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-820-339 Supplier Metro™ Supplier No. MXTTM24C
Add to Cart
Add to Cart
Includes
Posts, rigid casters, caster channels, bumpers and roller assemblies
High-density, easy access storage
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Mobile units move along anchored track allowing easy access to all shelves
Order Info
  • Order one per mobile unit
  • Shelves sold separately
Specifications
Product Type Mobile Unit Kit
Includes Posts, Chrome-plated rigid casters, caster channel, bumpers and roller assemblies
Width (Metric) Shelf 61 cm
For Use With (Equipment) Metro™ MetroMax i™ and Q™ Top-Track™ system
Width (English) Shelf 24 in.