Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ MetroMax™ iQ Polymer Shelving Accessory, Color Shelf Marker

Catalog No. 13451102 Shop All Metro Products
Change view
Click to view available options
Color:
Blue
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
13-451-102 Red
13-451-101 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-451-102 Supplier Metro™ Supplier No. CSM6RX
Add to Cart
Add to Cart

Use to assign color coding to individual shelf levels, shelving units, carts, or strorage areas.

  • Attaches easity to shelf for content identification
  • 6 in. length

Specifications

Product Type Shelf Marker
Color Red
For Use With (Equipment) Metro™ MetroMax i™ and Q™ shelving
Length (English) 6 in.
Length (Metric) 15.2 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.